Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024334.2 | NP_077310.1 | 8 | 916 | ||
XM_017007176.1 | XP_016862665.1 | 9 | 1038 | |||
GenBank mRNA | AK027466.1 | - | 2 | 146 | ||
AK027757.1 | - | 8 | 842 | |||
AK027827.1 | - | 3 | 199 | |||
AK027877.1 | - | 8 | 780 | |||
AK074073.1 | - | 6 | 452 | |||
AK075010.1 | - | 8 | 767 | |||
AK225589.1 | - | 8 | 839 | |||
AL136916.1 | - | 8 | 810 | |||
AY358625.1 | - | 8 | 679 | |||
BC003125.1 | - | 8 | 811 | 3503636 | ||
BC008054.2 | - | 6 | 406 | |||
BC011719.2 | - | 8 | 811 | 4079766 | ||
KU178769.1 | - | 7 | 662 | |||
KU178770.1 | - | 7 | 662 |