Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020989.3 | NP_066269.1 | 2 | 93 | ||
XM_011510662.1 | XP_011508964.1 | 3 | 167 | |||
GenBank mRNA | BC074954.2 | - | 2 | 93 | 30915279 | |
BC074955.2 | - | 2 | 93 | 30915501 | ||
KT455014.1 | - | 2 | 93 | |||
U66582.1 | - | 1 | 55 |