Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZXDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040653.3 | NP_001035743.1 | 2 | 1073 | ||
NM_025112.4 | NP_079388.3 | 2 | 1073 | |||
NR_104249.1 | 2 | 1073 | ||||
XM_005247757.3 | XP_005247814.1 | 2 | 1084 | |||
XM_006713741.2 | XP_006713804.1 | 2 | 1085 | |||
XM_011513119.2 | XP_011511421.1 | 2 | 1092 | |||
GenBank mRNA | AK092101.1 | - | 2 | 434 | ||
AY936556.1 | - | 2 | 1033 | |||
BC002940.2 | - | 11 | ||||
BC012729.2 | - | 11 | ||||
BC063713.1 | - | 2 | 141 | |||
DQ079590.1 | - | 2 | 1073 | |||
FJ980275.1 | - | 2 | 286 |