Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BHLHE41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030762.2 | NP_110389.1 | 2, 3 | 422 | ||
GenBank mRNA | AB044088.1 | - | 2, 3 | 265 | ||
AK303677.1 | - | 2, 3 | 261 | |||
AY665467.1 | - | 2, 3 | 232 | |||
BC025968.1 | - | 2, 3 | 422 | 4650325 |