Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286423.1 | NP_001273352.1 | 5 | 679 | ||
NM_001286427.1 | NP_001273356.1 | 4 | 466 | |||
NM_024506.4 | NP_078782.3 | 5 | 667 | |||
XM_011511813.2 | XP_011510115.1 | 4 | 431 | |||
XM_017004895.1 | XP_016860384.1 | 5 | 633 | |||
GenBank mRNA | AK055098.1 | - | 2 | 124 | ||
AK299404.1 | - | 4 | 428 | |||
AK300215.1 | - | 4 | 449 | |||
AY358639.1 | - | 5 | 647 |