Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001888.4 | NP_001879.1 | 6, 7 | 849 | ||
GenBank mRNA | AK290852.1 | - | 4, 5 | 568 | ||
BC018061.1 | - | 4, 5 | 567 | 4817634 | ||
BI860684.1 | - | 5, 6 | 522 | |||
BX648477.1 | - | 6, 7 | 765 | |||
L02950.1 | - | 4, 5 | 528 | |||
U85772.1 | - | 4, 5 | 523 |