Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPS8L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319952.1 | NP_001306881.1 | 14 | 1456 | ||
NM_024526.3 | NP_078802.2 | 15 | 1508 | |||
NM_133181.3 | NP_573444.2 | 15 | 1598 | |||
NM_139053.2 | NP_620641.1 | 15 | 1601 | |||
XM_011542132.1 | XP_011540434.1 | 15 | 1523 | |||
XM_011542133.1 | XP_011540435.1 | 15 | 1520 | |||
XM_011542134.2 | XP_011540436.1 | 14 | 1522 | |||
XM_011542135.2 | XP_011540437.1 | 15 | 1550 | |||
XM_017002327.1 | XP_016857816.1 | 15 | 1523 | |||
XM_017002328.1 | XP_016857817.1 | 15 | 1433 | |||
XM_017002329.1 | XP_016857818.1 | 15 | 1409 | |||
GenBank mRNA | AK025175.1 | - | 15 | 1410 | ||
AK225235.1 | - | 15 | 1410 | |||
AK290261.1 | - | 15 | 1435 | |||
AK292198.1 | - | 15 | 1522 | |||
AK307932.1 | - | 14 | 1354 | |||
AK308196.1 | - | 14 | 1339 | |||
AK310979.1 | - | 15 | 1495 | |||
AY074930.1 | - | 15 | 1484 | |||
BC012926.1 | - | 15 | 1521 | 3853503 | ||
BM824107.1 | - | 4 | 291 |