Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253826.1 | NP_001240755.1 | 5 | 1084 | ||
NM_001253827.1 | NP_001240756.1 | 6 | 849 | |||
NM_024572.3 | NP_078848.2 | 4 | 1069 | |||
XM_011533105.1 | XP_011531407.1 | 6 | 603 | |||
XM_017004906.1 | XP_016860395.1 | 4 | 534 | |||
XM_017004907.1 | XP_016860396.1 | 4 | 545 | |||
XM_017004908.1 | XP_016860397.1 | 4 | 828 | |||
GenBank mRNA | AB078144.1 | - | 4 | 429 | ||
AK022753.1 | - | 5 | 505 | |||
AK091313.1 | - | 4 | 439 | |||
AK122747.1 | - | 6 | 833 | |||
AK225808.1 | - | 4 | 793 | |||
AK298735.1 | - | 5 | 497 | |||
AK309417.1 | - | 6 | 688 | |||
AY358758.1 | - | 4 | 762 | |||
BC010659.1 | - | 4 | 1069 | 3874588 | ||
Y09324.4 | - | 4 | 429 |