Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024578.2 | NP_078854.1 | 6 | 770 | ||
XM_005260079.2 | XP_005260136.1 | 6 | 716 | |||
GenBank mRNA | AF109191.1 | - | 4 | 309 | ||
AK026362.1 | - | 6 | 728 | |||
BC004121.2 | - | 5 | 982 | |||
BC029361.1 | - | 6 | 770 | 4064124 |