Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146075.1 | NP_001139547.1 | 13 | 1590 | ||
NM_001146076.1 | NP_001139548.1 | 13 | 1533 | |||
NM_024604.2 | NP_078880.2 | 14 | 1692 | |||
XM_011538741.2 | XP_011537043.1 | 12 | 1443 | |||
GenBank mRNA | AK025561.1 | - | 14 | 1622 | ||
AK299465.1 | - | 13 | 1498 | |||
BC056415.1 | - | 13 | 1515 | 5218249 |