Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302084.1 | NP_001289013.1 | 10 | 1076 | ||
NM_024650.3 | NP_078926.3 | 12 | 1365 | |||
GenBank mRNA | AK026184.1 | - | 11 | 1166 | ||
AK225435.1 | - | 11 | 1166 | |||
AK302025.1 | - | 10 | 1028 | |||
BC028240.1 | - | 12 | 1368 | 5243907 |