Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001085354.2 | NP_001078823.1 | 3 | 489 | ||
NM_024657.4 | NP_078933.3 | 3 | 489 | |||
XM_005262190.3 | XP_005262247.1 | 2 | 379 | |||
XM_006724691.2 | XP_006724754.1 | 3 | 489 | |||
XM_017029844.1 | XP_016885333.1 | 2 | 379 | |||
GenBank mRNA | AK300305.1 | - | 2 | 64 | ||
AY927537.1 | - | LIT3543 AY927537 |
1 | 24 | ||
CB989767.1 | - | 3 | 484 | |||
DA713534.1 | - | 3 | 490 | |||
EF125209.1 | - | 3 | 214 | |||
EF125210.1 | - | 3 | 496 |