Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317776.1 | NP_001304705.1 | 6 | 2582 | ||
NM_024672.5 | NP_078948.3 | 5 | 2516 | |||
XM_005263220.2 | XP_005263277.1 | 5 | 2473 | |||
XM_011532246.2 | XP_011530548.1 | 5 | 2306 | |||
XM_017008603.1 | XP_016864092.1 | 5 | 3356 | |||
XM_017008604.1 | XP_016864093.1 | 5 | 3409 | |||
XM_017008605.1 | XP_016864094.1 | 5 | 2450 | |||
XM_017008606.1 | XP_016864095.1 | 6 | 2574 | |||
XM_017008607.1 | XP_016864096.1 | 5 | 2789 | |||
XM_017008608.1 | XP_016864097.1 | 6 | 3428 | |||
XM_017008609.1 | XP_016864098.1 | 6 | 2578 | |||
XM_017008610.1 | XP_016864099.1 | 6 | 2565 | |||
GenBank mRNA | AK026973.1 | - | 3 | 1846 | ||
AK091412.1 | - | 5 | 2413 | |||
AK225333.1 | - | 3 | 1853 | |||
BC131722.1 | - | 6 | 2463 | |||
BX648134.1 | - | 6 | 2736 |