Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324171.1 | NP_001311100.1 | 10 | 874 | ||
NM_001324172.1 | NP_001311101.1 | 10 | 874 | |||
NM_030581.3 | NP_085058.3 | 10 | 847 | |||
XM_005256146.3 | XP_005256203.1 | 10 | 851 | |||
XM_011523331.2 | XP_011521633.1 | 9 | 691 | |||
XM_011523332.2 | XP_011521634.1 | 4 | 274 | |||
XM_017023667.1 | XP_016879156.1 | 9 | 708 | |||
XM_017023668.1 | XP_016879157.1 | 8 | 484 | |||
XM_017023669.1 | XP_016879158.1 | 9 | 579 | |||
XM_017023670.1 | XP_016879159.1 | 8 | 587 | |||
GenBank mRNA | AB067510.1 | - | 5 | 320 | ||
AK302557.1 | - | 10 | 915 | |||
BC004519.2 | - | 10 | 811 | 3938206 | ||
BC014887.2 | - | 4 | 239 |