Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024686.4 | NP_078962.4 | 19 | 2708 | ||
XM_005271208.2 | XP_005271265.1 | 18 | 2519 | |||
XM_017002347.1 | XP_016857836.1 | 20 | 2728 | |||
XM_017002348.1 | XP_016857837.1 | 19 | 2898 | |||
XM_017002349.1 | XP_016857838.1 | 18 | 2483 | |||
GenBank mRNA | AK026686.1 | - | 12 | 1474 | ||
AK094174.1 | - | 14 | 2370 | |||
AK126792.1 | - | 19 | 2605 | |||
AK128604.1 | - | 16 | 2683 | |||
AK225574.1 | - | 12 | 1474 | |||
AY170843.1 | - | 18 | 2542 | |||
BC028350.2 | - | 12 | 1644 | |||
BC032829.1 | - | 19 | 2557 | |||
BC048970.1 | - | 22 | 2905 | 5295193 | ||
BC060878.1 | - | 19 | 2690 | 5270163 |