Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025105.2 | NP_001020276.1 | 3, 4 | 953 | ||
NM_001271741.1 | NP_001258670.1 | 3, 4 | 953 | |||
NM_001271742.1 | NP_001258671.1 | 2, 3 | 392 | |||
NM_001892.5 | NP_001883.4 | 3, 4 | 953 | |||
GenBank mRNA | AF218004.1 | - | 3, 4 | 751 | ||
AK293761.1 | - | 2, 3 | 359 | |||
AK294942.1 | - | 4, 5 | 540 | |||
BC007246.1 | - | 3, 4 | 774 | |||
BC008717.2 | - | 3, 4 | 495 | 2822250 | ||
BC021971.2 | - | 3, 4 | 772 | |||
BC025371.2 | - | 3, 4 | 772 | |||
BT019829.1 | - | 3, 4 | 365 | |||
BT019830.1 | - | 3, 4 | 365 | |||
CR542206.1 | - | 3, 4 | 365 | |||
DQ082865.1 | - | 3, 4 | 365 | |||
EU668329.1 | - | 3, 4 | 365 | |||
L37042.1 | - | 3, 4 | 576 | |||
X80693.1 | - | 3, 4 | 490 |
Gene Symbol: CSNK1A1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027320.1 | 1 | 727 | |||
GenBank mRNA | BC028192.1 | - | 1 | 680 |