Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD4 SIRNA Assays ›
Gene Symbol:
ACBD4
Gene Name:
acyl-CoA binding domain containing 4
Gene Aliases:
HMFT0700
Chromosome Location:
Chr.17: 45132600 - 45144181 on Build GRCh38
UniGene ID:
Hs.110298
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36304
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135705.2
NP_001129177.1
4
618
NM_001135706.2
NP_001129178.1
4
618
NM_001135707.2
NP_001129179.1
4
618
NM_001321352.1
NP_001308281.1
6
742
NM_001321353.1
NP_001308282.1
6
739
NM_024722.3
NP_078998.1
3
772
XM_006722085.2
XP_006722148.1
5
988
XM_011525259.2
XP_011523561.1
5
988
XM_011525261.2
XP_011523563.1
5
988
XM_017025084.1
XP_016880573.1
5
988
XM_017025085.1
XP_016880574.1
6
739
XM_017025086.1
XP_016880575.1
7
888
XM_017025087.1
XP_016880576.1
5
988
XM_017025088.1
XP_016880577.1
5
988
XM_017025089.1
XP_016880578.1
5
988
XM_017025090.1
XP_016880579.1
5
988
XM_017025091.1
XP_016880580.1
5
988
XM_017025092.1
XP_016880581.1
6
1113
XM_017025093.1
XP_016880582.1
5
988
XM_017025094.1
XP_016880583.1
3
732
XM_017025095.1
XP_016880584.1
5
988
XM_017025096.1
XP_016880585.1
5
988
XM_017025097.1
XP_016880586.1
5
988
XM_017025098.1
XP_016880587.1
6
739
XM_017025099.1
XP_016880588.1
5
988
XM_017025100.1
XP_016880589.1
5
988
XM_017025101.1
XP_016880590.1
5
988
GenBank mRNA
AB073387.1
-
4
530
AK023384.1
-
3
732
AK075104.1
-
4
571
AK296167.1
-
4
605
AK316359.1
-
4
618
BC029164.1
-
4
557
5188183
BC041143.1
-
6
711
5769682
KU178779.1
-
3
274
KU178780.1
-
3
274
KU178781.1
-
3
274
KU178782.1
-
3
274
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Process(es)
fatty-acyl-CoA binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0