Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270584.1 | NP_001257513.1 | 6 | 1122 | ||
NM_001270585.1 | NP_001257514.1 | 6 | 910 | |||
NM_024726.4 | NP_079002.3 | 6 | 1122 | |||
NR_073043.1 | 6 | 1122 | ||||
XM_011511864.1 | XP_011510166.1 | 6 | 908 | |||
XM_011511865.2 | XP_011510167.1 | 6 | 1145 | |||
XM_011511866.1 | XP_011510168.1 | 6 | 908 | |||
XM_017004960.1 | XP_016860449.1 | 6 | 908 | |||
GenBank mRNA | AK054711.1 | - | 6 | 909 | ||
AK294105.1 | - | 6 | 1045 | |||
AK295566.1 | - | 6 | 907 | |||
BC028699.1 | - | 6 | 1125 | 4839325 | ||
BC043504.1 | - | 6 | 1036 | 5193742 |