Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193311.1 | NP_001180240.1 | 11 | 947 | ||
NM_001193312.1 | NP_001180241.1 | 10 | 803 | |||
NM_001193313.1 | NP_001180242.1 | 11 | 947 | |||
NM_024728.2 | NP_079004.1 | 11 | 836 | |||
XM_006715775.3 | XP_006715838.1 | 11 | 1036 | |||
XM_011515525.2 | XP_011513827.1 | 11 | 943 | |||
XM_011515526.2 | XP_011513828.1 | 11 | 943 | |||
XM_011515527.2 | XP_011513829.1 | 10 | 799 | |||
XM_011515528.2 | XP_011513830.1 | 11 | 943 | |||
XM_011515529.2 | XP_011513831.1 | 10 | 799 | |||
XM_017012621.1 | XP_016868110.1 | 11 | 1052 | |||
XM_017012622.1 | XP_016868111.1 | 11 | 943 | |||
GenBank mRNA | AB014767.1 | - | 10 | 800 | ||
AK021870.1 | - | 11 | 836 | |||
AK296048.1 | - | 11 | 914 | |||
AK299133.1 | - | 11 | 927 | |||
AK307935.1 | - | 11 | 906 | |||
AK308764.1 | - | 11 | 907 | |||
BC098117.1 | - | 11 | 946 | 40024552 | ||
BC098261.1 | - | 11 | 946 | 40024553 | ||
BC098310.1 | - | 10 | 802 | 40024556 | ||
BC098318.1 | - | 11 | 946 | 40024555 |