Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSNK1G3 SIRNA Assays ›
Gene Symbol:
CSNK1G3
Gene Name:
casein kinase 1 gamma 3
Gene Aliases:
CKI-gamma 3, CSNK1G3L
Chromosome Location:
Chr.5: 123512099 - 123617045 on Build GRCh38
UniGene ID:
Hs.129206
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s3635
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSNK1G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031812.3
NP_001026982.1
9
1653
NM_001044723.2
NP_001038188.1
8
1181
NM_001270572.1
NP_001257501.1
8
1184
NM_001270573.1
NP_001257502.1
8
1228
NM_001270574.1
NP_001257503.1
6
1120
NM_004384.4
NP_004375.2
8
1181
XM_005271891.1
XP_005271948.1
9
1656
XM_005271892.2
XP_005271949.1
9
1653
XM_005271894.2
XP_005271951.1
9
1653
XM_005271895.2
XP_005271952.1
9
1656
XM_005271896.1
XP_005271953.1
9
1656
XM_017009057.1
XP_016864546.1
10
1731
XM_017009058.1
XP_016864547.1
12
1940
XM_017009059.1
XP_016864548.1
11
1799
XM_017009060.1
XP_016864549.1
11
1855
XM_017009061.1
XP_016864550.1
10
1728
XM_017009062.1
XP_016864551.1
10
1287
XM_017009063.1
XP_016864552.1
10
1731
XM_017009064.1
XP_016864553.1
10
1731
XM_017009065.1
XP_016864554.1
10
1728
XM_017009066.1
XP_016864555.1
11
1796
XM_017009067.1
XP_016864556.1
10
1731
XM_017009068.1
XP_016864557.1
10
1728
XM_017009069.1
XP_016864558.1
10
1724
XM_017009070.1
XP_016864559.1
9
1653
GenBank mRNA
AF049089.1
-
8
964
AF049090.1
-
8
964
AK289405.1
-
8
1129
AK299739.1
-
8
1067
AK316019.1
-
6
959
BC047567.1
-
9
1305
4817058
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_152809, BC033601, AK032675, AK218781.
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
endocytosis
signal transduction
regulation of cell shape
Wnt signaling pathway
peptidyl-serine phosphorylation
protein kinase activity
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0