Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HHIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024746.3 | NP_079022.2 | 2 | 597 | ||
XM_011509985.2 | XP_011508287.1 | 2 | 542 | |||
XM_011509986.1 | XP_011508288.1 | 2 | 542 | |||
XM_017002350.1 | XP_016857839.1 | 2 | 542 | |||
GenBank mRNA | AK023902.1 | - | 2 | 282 | ||
AY358602.1 | - | 2 | 579 | |||
BC007638.1 | - | 2 | 583 | 3533045 | ||
BC015201.1 | - | 1 | 188 | |||
BC016552.1 | - | 2 | 597 | 4452939 |