Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAND2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136192.1 | 4 | 1300 | |||
NR_136193.1 | 4 | 1997 | ||||
NR_136194.1 | 4 | 1467 | ||||
NR_136197.1 | 4 | 1371 | ||||
NR_136198.1 | 3 | 1188 | ||||
NR_136199.1 | 5 | 1510 | ||||
NR_136200.1 | 4 | 2000 | ||||
NR_136201.1 | 4 | 2167 | ||||
GenBank mRNA | AB075501.1 | - | 4 | 1908 | ||
AK021601.1 | - | 1 | 1647 | |||
DQ831955.1 | - | 1 | 60 | |||
DQ831956.1 | - | 1 | 60 | |||
EF529498.1 | - | 4 | 435 | |||
EU000853.1 | - | 4 | 1213 | |||
EU000854.1 | - | 4 | 1213 | |||
EU000855.1 | - | 3 | 2305 | |||
EU000856.1 | - | 3 | 2305 |