Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VASH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136474.2 | NP_001129946.1 | 7 | 971 | ||
NM_001136475.2 | NP_001129947.1 | 5 | 687 | |||
NM_001301056.1 | NP_001287985.1 | 6 | 1167 | |||
NM_024749.4 | NP_079025.2 | 4 | 1035 | |||
XM_011509987.2 | XP_011508289.1 | 9 | 1451 | |||
XM_011509988.2 | XP_011508290.1 | 10 | 1525 | |||
XM_011509989.2 | XP_011508291.1 | 6 | 1145 | |||
XM_011509991.2 | XP_011508293.2 | 5 | 864 | |||
XM_011509995.1 | XP_011508297.1 | 4 | 567 | |||
XM_017002351.1 | XP_016857840.1 | 7 | 1285 | |||
XM_017002352.1 | XP_016857841.1 | 4 | 718 | |||
GenBank mRNA | AK022567.1 | - | 7 | 897 | ||
AK096605.1 | - | 7 | 835 | |||
AK290973.1 | - | 7 | 930 | |||
AK302675.1 | - | 5 | 540 | |||
AY834202.1 | - | 5 | 742 | |||
BC028194.1 | - | 4 | 615 | 5164251 | ||
BC051856.1 | - | 4 | 920 | 6067012 |