Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024761.4 | NP_079037.3 | 3 | 1062 | ||
GenBank mRNA | AJ580636.1 | - | 2 | 566 | ||
AK023266.1 | - | 3 | 991 | |||
AL832572.2 | - | 3 | 956 | |||
BC033027.1 | - | 3 | 974 | 5268356 | ||
CR457307.1 | - | 3 | 566 |