Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003009.2 | NP_003000.1 | 6 | 848 | ||
GenBank mRNA | AF015283.1 | - | 6 | 697 | ||
AK026717.1 | - | 5 | 2410 | |||
AK091604.1 | - | 1 | 2476 | |||
BC000581.2 | - | 6 | 713 | 3162433 | ||
BC032546.1 | - | 6 | 738 | 5535970 | ||
BC039597.1 | - | 6 | 716 | 6015153 | ||
BC047893.1 | - | 6 | 715 | 6204763 | ||
U67171.1 | - | 6 | 708 |