Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300800.1 | NP_001287729.1 | 7 | 713 | ||
NM_024771.3 | NP_079047.2 | 7 | 642 | |||
XM_006718689.3 | XP_006718752.1 | 7 | 539 | |||
XM_011545254.2 | XP_011543556.1 | 8 | 716 | |||
XM_011545255.2 | XP_011543557.1 | 8 | 626 | |||
GenBank mRNA | AK023910.1 | - | 7 | 611 | ||
AK299041.1 | - | 7 | 707 | |||
AK300451.1 | - | 6 | 563 | |||
AK303557.1 | - | 5 | 473 | |||
BC041617.1 | - | 7 | 571 | 5921480 | ||
BC052298.1 | - | 7 | 571 | 6292760 | ||
BX647309.1 | - | 6 | 1668 | |||
JF263566.1 | - | 7 | 611 |