Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105248.1 | NP_001098718.1 | 7, 8 | 1992 | ||
NM_001105249.1 | NP_001098719.1 | 7, 8 | 1992 | |||
NM_001261841.1 | NP_001248770.1 | 7, 8 | 1947 | |||
NM_001308161.1 | NP_001295090.1 | 7, 8 | 1785 | |||
NM_024780.4 | NP_079056.2 | 3, 4 | 762 | |||
GenBank mRNA | AK097718.1 | - | 3, 4 | 727 | ||
AK131202.1 | - | 4, 5 | 482 | |||
AK299055.1 | - | 7, 8 | 1785 | |||
AK304431.1 | - | 7, 8 | 1947 | |||
AY236494.1 | - | 3, 4 | 762 | |||
AY263164.1 | - | 3, 4 | 505 | |||
AY358155.1 | - | 5, 6 | 1341 | |||
BC027602.1 | - | 3, 4 | 717 | 4839111 | ||
BC038118.2 | - | 3, 4 | 457 | 5265527 | ||
CR749359.1 | - | 5, 6 | 647 |