Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143850.2 | NP_001137322.1 | 15 | 1857 | ||
NM_024809.4 | NP_079085.2 | 15 | 1860 | |||
XM_005253623.2 | XP_005253680.1 | 14 | 1725 | |||
XM_006719605.3 | XP_006719668.1 | 15 | 1860 | |||
XM_017019974.1 | XP_016875463.1 | 14 | 1722 | |||
XM_017019975.1 | XP_016875464.1 | 14 | 1735 | |||
GenBank mRNA | AK023037.1 | - | 4 | 416 | ||
AK056924.1 | - | 15 | 1858 | |||
AK292153.1 | - | 15 | 1857 | |||
BC009112.1 | - | 15 | 1829 | 3463475 | ||
DQ278870.1 | - | 15 | 1858 |