Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146254.1 | NP_001139726.1 | 2 | 470 | ||
NM_001146255.1 | NP_001139727.1 | 2 | 470 | |||
NM_024825.3 | NP_079101.3 | 2 | 494 | |||
XM_006722901.1 | XP_006722964.1 | 2 | 427 | |||
XM_006722902.1 | XP_006722965.1 | 2 | 427 | |||
XM_006722903.1 | XP_006722966.1 | 2 | 427 | |||
XM_011528308.1 | XP_011526610.1 | 2 | 461 | |||
XM_011528309.2 | XP_011526611.1 | 2 | 440 | |||
XM_011528310.2 | XP_011526612.1 | 2 | 439 | |||
XM_011528311.1 | XP_011526613.1 | 2 | 427 | |||
XM_011528313.1 | XP_011526615.1 | 2 | 461 | |||
XM_011528314.1 | XP_011526616.1 | 2 | 427 | |||
XM_011528316.1 | XP_011526618.1 | 2 | 461 | |||
XM_017027313.1 | XP_016882802.1 | 2 | 461 | |||
GenBank mRNA | AK027100.1 | - | 2 | 460 | ||
AK290731.1 | - | 2 | 427 | |||
AK296101.1 | - | 1 | 255 | |||
AK298489.1 | - | 2 | 470 | |||
BC057786.1 | - | 2 | 494 | 30347370 |