Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024857.4 | NP_079133.3 | 2 | 850 | ||
XM_011525269.2 | XP_011523571.1 | 2 | 871 | |||
XM_011525270.2 | XP_011523572.1 | 2 | 871 | |||
XM_011525273.2 | XP_011523575.1 | 2 | 1888 | |||
XM_017025111.1 | XP_016880600.1 | 2 | 846 | |||
GenBank mRNA | AJ314648.2 | - | 2 | 818 | ||
AL832103.1 | - | 2 | 851 | |||
AY557611.1 | - | 2 | 818 | |||
BC015051.1 | - | 2 | 839 |