Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGIX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005278064.4 | XP_005278121.1 | 2 | 265 | ||
XM_005278065.3 | XP_005278122.1 | 2 | 353 | |||
XM_011543985.2 | XP_011542287.1 | 3 | 486 | |||
XM_011543986.2 | XP_011542288.1 | 2 | 240 | |||
XM_011543987.2 | XP_011542289.1 | 2 | 261 | |||
GenBank mRNA | AK025340.1 | - | 2 | 187 | ||
AK096058.1 | - | 2 | 1427 | |||
DQ884401.1 | - | 1 | 108 |