Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085437.2 | NP_001078906.2 | 5 | 1424 | ||
NM_001282921.1 | NP_001269850.1 | 5 | 917 | |||
NM_024861.3 | NP_079137.3 | 5 | 860 | |||
XM_011511877.1 | XP_011510179.1 | 6 | 1416 | |||
GenBank mRNA | AK026324.1 | - | 5 | 857 | ||
AK056601.1 | - | 5 | 1424 | |||
AK096434.1 | - | 5 | 917 | |||
BC125154.1 | - | 5 | 848 | 40120634 | ||
BC125155.1 | - | 5 | 848 | 40120635 |