Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006935.2 | NP_001006936.1 | 3 | 421 | ||
NM_001006937.2 | NP_001006938.1 | 3 | 416 | |||
NM_001300901.1 | NP_001287830.1 | 5 | 849 | |||
NM_001305840.1 | NP_001292769.1 | 4 | 670 | |||
NM_001305841.1 | NP_001292770.1 | 2 | 343 | |||
NM_001305842.1 | NP_001292771.1 | 3 | 475 | |||
NM_024863.5 | NP_079139.4 | 3 | 480 | |||
XM_017029856.1 | XP_016885345.1 | 3 | 740 | |||
XM_017029857.1 | XP_016885346.1 | 3 | 751 | |||
GenBank mRNA | AF271783.1 | - | 3 | 393 | ||
AF314542.1 | - | 5 | 849 | |||
AK024827.1 | - | 3 | 414 | |||
AK093021.1 | - | 1 | 1347 | |||
AK225117.1 | - | 3 | 414 | |||
AL833640.1 | - | 3 | 474 | |||
AY885169.1 | - | 3, 4 | 384 | |||
BC012296.1 | - | 3 | 380 | 3445403 | ||
BI084326.1 | - | 2 | 313 | |||
BP375855.1 | - | 3 | 409 | |||
DA230626.1 | - | 3 | 416 |