Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNTNAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033655.3 | NP_387504.2 | 12 | 2048 | ||
GenBank mRNA | AB051501.2 | - | 12 | 1955 | ||
AF333769.2 | - | 12 | 1916 | |||
AK056833.1 | - | 12 | 2048 | |||
AK294281.1 | - | 10 | 1561 | |||
BC132737.1 | - | 12 | 1953 | 40146759 | ||
BC144130.1 | - | 10 | 1674 |