Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261392.1 | NP_001248321.1 | 19 | 1942 | ||
NM_001261394.1 | NP_001248323.1 | 19 | 1870 | |||
NM_024894.3 | NP_079170.2 | 20 | 2020 | |||
NR_048552.1 | 19 | 1974 | ||||
XM_011510398.2 | XP_011508700.1 | 19 | 1887 | |||
XM_011510399.2 | XP_011508701.1 | 18 | 1825 | |||
XM_017004990.1 | XP_016860479.1 | 16 | 1545 | |||
XM_017004991.1 | XP_016860480.1 | 9 | 997 | |||
GenBank mRNA | AK024137.1 | - | 19 | 1855 | ||
AK290680.1 | - | 20 | 1984 | |||
AK297169.1 | - | 19 | 1930 | |||
AK299617.1 | - | 19 | 1962 | |||
AL529108.3 | - | 1 | 42 | |||
BC005125.2 | - | 20 | 2000 | 3841643 | ||
DN602355.1 | - | 5 | 648 |