Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024899.3 | NP_079175.2 | 4 | 615 | ||
NR_073537.1 | 4 | 610 | ||||
XM_005258149.4 | XP_005258206.1 | 4 | 594 | |||
XM_017025981.1 | XP_016881470.1 | 4 | 594 | |||
XM_017025982.1 | XP_016881471.1 | 4 | 675 | |||
XM_017025983.1 | XP_016881472.1 | 4 | 594 | |||
XM_017025984.1 | XP_016881473.1 | 4 | 577 | |||
XM_017025985.1 | XP_016881474.1 | 4 | 543 | |||
XM_017025986.1 | XP_016881475.1 | 3 | 352 | |||
XM_017025987.1 | XP_016881476.1 | 3 | 319 | |||
GenBank mRNA | AK022604.1 | - | 4 | 477 | ||
AK298223.1 | - | 4 | 577 | |||
AK301964.1 | - | 3 | 352 | |||
AL833727.1 | - | 4 | 463 | |||
BC026307.1 | - | 4 | 542 | 4792211 | ||
BC033026.1 | - | 4 | 477 |