Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287437.1 | NP_001274366.1 | 4 | 481 | ||
NM_001287439.1 | NP_001274368.1 | 4 | 599 | |||
NM_001287440.1 | NP_001274369.1 | 4 | 396 | |||
NM_001287441.1 | NP_001274370.1 | 4 | 360 | |||
NM_001287442.1 | NP_001274371.1 | 4 | 353 | |||
NM_001287443.1 | NP_001274372.1 | 4 | 357 | |||
NM_024900.4 | NP_079176.2 | 4 | 481 | |||
NM_199320.3 | NP_955352.1 | 4 | 481 | |||
XM_005263232.1 | XP_005263289.1 | 4 | 323 | |||
XM_017008626.1 | XP_016864115.1 | 4 | 438 | |||
XM_017008627.1 | XP_016864116.1 | 4 | 485 | |||
GenBank mRNA | AF520952.1 | - | 4 | 378 | ||
AK027620.1 | - | 4 | 401 | |||
AK127326.1 | - | 4 | 289 | |||
AK304235.1 | - | 4 | 314 | |||
AK304815.1 | - | 4 | 295 | |||
BC032376.1 | - | 4 | 315 | 5111727 | ||
BI463062.1 | - | 4 | 499 | |||
DA478923.1 | - | 4 | 395 | |||
DA714593.1 | - | 4 | 353 | |||
DB085413.1 | - | 4 | 357 | |||
DB293142.1 | - | 4 | 335 | |||
DB451567.1 | - | 4 | 480 |