Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323.3 | NP_001314.1 | 3 | 578 | ||
GenBank mRNA | AK092391.1 | - | 1 | 1985 | ||
BC031334.1 | - | 3 | 549 | 5446833 | ||
U62800.1 | - | 3 | 547 | |||
U81233.1 | - | 3 | 542 |