Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024954.4 | NP_079230.1 | 2 | 473 | ||
GenBank mRNA | AK021869.1 | - | 2 | 319 | ||
AK222597.1 | - | 2 | 342 | |||
BC007331.2 | - | 2 | 344 | 3629185 | ||
BM922594.1 | - | 2 | 482 |