Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323679.1 | NP_001310608.1 | 6 | 2027 | ||
NM_001323680.1 | NP_001310609.1 | 4 | 2108 | |||
NM_001323681.1 | NP_001310610.1 | 4 | 2099 | |||
NM_001323682.1 | NP_001310611.1 | 5 | 2165 | |||
NM_001323683.1 | NP_001310612.1 | 5 | 2159 | |||
NM_001323684.1 | NP_001310613.1 | 5 | 2187 | |||
NM_001323685.1 | NP_001310614.1 | 4 | 1828 | |||
NM_024958.2 | NP_079234.1 | 4 | 2086 | |||
NR_136649.1 | 3 | 1891 | ||||
XM_006723630.3 | XP_006723693.1 | 4 | 2509 | |||
XM_011529360.1 | XP_011527662.1 | 4 | 1792 | |||
XM_011529362.2 | XP_011527664.1 | 5 | 1799 | |||
XM_011529363.2 | XP_011527665.1 | 4 | 1777 | |||
XM_017028073.1 | XP_016883562.1 | 4 | 1731 | |||
XM_017028074.1 | XP_016883563.1 | 4 | 1785 | |||
XM_017028075.1 | XP_016883564.1 | 4 | 1957 | |||
GenBank mRNA | AK026982.1 | - | 4 | 1696 | ||
AL136915.1 | - | 3 | 1665 | |||
BC001963.1 | - | 3 | 1665 | 3454427 |