Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF7IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256160.1 | NP_001243089.1 | 4 | 822 | ||
NM_024997.3 | NP_079273.2 | 3 | 779 | |||
NR_045815.1 | 2 | 711 | ||||
NR_045816.1 | 3 | 779 | ||||
XM_006720953.3 | XP_006721016.1 | 2 | 606 | |||
XM_006720954.3 | XP_006721017.1 | 2 | 1745 | |||
XM_017023705.1 | XP_016879194.1 | 2 | 1747 | |||
XM_017023706.1 | XP_016879195.1 | 2 | 1747 | |||
XM_017023707.1 | XP_016879196.1 | 2 | 1747 | |||
XM_017023710.1 | XP_016879199.1 | 1 | 313 | |||
GenBank mRNA | AK093414.1 | - | 4 | 820 | ||
AK299040.1 | - | 3 | 770 | |||
AL832987.1 | - | 1 | 501 | |||
AY560615.1 | - | 1 | 549 | |||
BC033891.1 | - | 3 | 775 | 5266089 | ||
BC137079.1 | - | 2 | 678 | 9021076 | ||
BC144468.1 | - | 2 | 678 |