Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025074.6 | NP_079350.5 | 55 | 8334 | ||
XM_006714316.2 | XP_006714379.1 | 54 | 8353 | |||
GenBank mRNA | AB040933.2 | - | 17 | 2579 | ||
AK092082.1 | - | 2 | 211 | |||
BX647949.1 | - | 5 | 614 |