Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025082.3 | NP_079358.3 | 14, 15 | 2004 | ||
XM_006721277.2 | XP_006721340.1 | 12, 13 | 1859 | |||
XM_011523344.1 | XP_011521646.1 | 13, 14 | 1981 | |||
XM_011523345.1 | XP_011521647.1 | 5, 6 | 982 | |||
XM_017023712.1 | XP_016879201.1 | 12, 13 | 1858 | |||
XM_017023713.1 | XP_016879202.1 | 12, 13 | 2286 | |||
XM_017023714.1 | XP_016879203.1 | 12, 13 | 2391 | |||
XM_017023715.1 | XP_016879204.1 | 9, 10 | 1731 | |||
XM_017023716.1 | XP_016879205.1 | 12, 13 | 2652 | |||
XM_017023717.1 | XP_016879206.1 | 5, 6 | 1094 | |||
GenBank mRNA | AK023173.1 | - | 10, 11 | 1794 | ||
AK056097.1 | - | 14, 15 | 2004 | |||
AK225399.1 | - | 11, 12 | 1493 | |||
AK291527.1 | - | 11, 12 | 1524 | |||
AK301621.1 | - | 11, 12 | 1407 | |||
BC007642.1 | - | 11, 12 | 1447 | |||
BC007864.2 | - | 2, 3 | 253 | |||
BC015202.2 | - | 11, 12 | 1455 | 3878214 | ||
BX648617.1 | - | 7, 8 | 1434 | |||
CD242240.1 | - | 1, 2 | 95 |