Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025108.2 | NP_079384.2 | 4 | 297 | ||
XM_011522667.1 | XP_011520969.1 | 3 | 201 | |||
GenBank mRNA | AK023971.1 | - | 3 | 378 | ||
AK093788.1 | - | 3 | 535 | |||
AK301929.1 | - | 4 | 293 | |||
AL559561.3 | - | 4 | 297 | |||
BC018719.1 | - | 4 | 285 | 4868952 |