Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 8 | 1652 | ||
NM_001286762.1 | NP_001273691.1 | 7 | 1558 | |||
NM_001286763.1 | NP_001273692.1 | 8 | 1287 | |||
NM_001286764.1 | NP_001273693.1 | 7 | 1171 | |||
NM_001286765.1 | NP_001273694.1 | 7 | 1017 | |||
NM_001286766.1 | NP_001273695.1 | 6 | 901 | |||
NM_025113.3 | NP_079389.2 | 8 | 1674 | |||
GenBank mRNA | AK025215.1 | - | 7 | 1307 | ||
AK093073.1 | - | 8 | 1622 | |||
AK125950.1 | - | 8 | 1659 | |||
AK293034.1 | - | 7 | 923 | |||
AK300183.1 | - | 7 | 721 | |||
AK307854.1 | - | 6 | 644 | |||
AK310264.1 | - | 9 | 1661 | |||
BC004495.1 | - | 4 | 371 | |||
BC032311.1 | - | 6 | 1178 | |||
BC043488.1 | - | 8 | 1020 | 5205680 | ||
BX641126.1 | - | 6 | 708 |