Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0226L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286761.1 | NP_001273690.1 | 11 | 1946 | ||
NM_001286762.1 | NP_001273691.1 | 10 | 1852 | |||
NM_001286763.1 | NP_001273692.1 | 11 | 1581 | |||
NM_001286764.1 | NP_001273693.1 | 10 | 1465 | |||
NM_001286765.1 | NP_001273694.1 | 10 | 1311 | |||
NM_001286766.1 | NP_001273695.1 | 9 | 1195 | |||
NM_025113.3 | NP_079389.2 | 11 | 1968 | |||
GenBank mRNA | AK025215.1 | - | 10 | 1601 | ||
AK093073.1 | - | 11 | 1916 | |||
AK125750.1 | - | 1 | 689 | |||
AK125950.1 | - | 11 | 1953 | |||
AK293034.1 | - | 10 | 1217 | |||
AK300183.1 | - | 10 | 1015 | |||
AK307854.1 | - | 9 | 938 | |||
AK310264.1 | - | 13 | 2056 | |||
BC004495.1 | - | 7 | 665 | |||
BC032311.1 | - | 9 | 2995 | |||
BC043488.1 | - | 11 | 1314 | 5205680 | ||
BX641126.1 | - | 9 | 1002 |