Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025129.4 | NP_079405.2 | 3 | 449 | ||
XM_006723399.3 | XP_006723462.1 | 3 | 427 | |||
XM_011527339.1 | XP_011525641.1 | 3 | 427 | |||
XM_011527340.1 | XP_011525642.1 | 3 | 289 | |||
XM_011527341.2 | XP_011525643.1 | 3 | 353 | |||
XM_011527342.1 | XP_011525644.1 | 3 | 576 | |||
XM_011527343.1 | XP_011525645.1 | 3 | 427 | |||
XM_011527345.1 | XP_011525647.1 | 3 | 385 | |||
XM_011527346.1 | XP_011525648.1 | 2 | 303 | |||
XM_011527347.1 | XP_011525649.1 | 4 | 343 | |||
XM_017027319.1 | XP_016882808.1 | 3 | 289 | |||
XM_017027320.1 | XP_016882809.1 | 3 | 427 | |||
XM_017027321.1 | XP_016882810.1 | 2 | 303 | |||
XM_017027322.1 | XP_016882811.1 | 3 | 289 | |||
XM_017027323.1 | XP_016882812.1 | 3 | 427 | |||
GenBank mRNA | AK026341.1 | - | 3 | 423 | ||
AK295072.1 | - | 3 | 448 | |||
AK315445.1 | - | 3 | 427 | |||
BC004445.1 | - | 3 | 350 | 2819805 | ||
BC016793.2 | - | 3 | 350 | 2819805 | ||
CB152506.1 | - | 2 | 327 |