Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002814.2 | NP_001002814.2 | 6 | 3811 | ||
NM_025151.4 | NP_079427.4 | 5 | 1909 | |||
GenBank mRNA | AF368294.1 | - | 5 | 1879 | ||
AK026177.1 | - | 2 | 150 | |||
AK092296.1 | - | 4 | 1217 | |||
AK122583.1 | - | 3 | 1950 | |||
AK124422.1 | - | 1 | 709 | |||
AK226111.1 | - | 6 | 3501 | |||
AK291781.1 | - | 5 | 1906 | |||
AL832155.1 | - | 3 | 1392 | |||
AL833169.1 | - | 3 | 1220 | |||
AY280966.1 | - | 3 | 2232 | |||
AY280968.1 | - | 6 | 3768 | |||
AY280969.1 | - | 2 | 2981 | |||
BC077720.1 | - | 5 | 1879 | 6537110 | ||
DQ083995.1 | - | 5 | 1633 | |||
DQ236342.1 | - | 5 | 1879 |