Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320484.1 | NP_001307413.1 | 10 | 1827 | ||
NM_001320485.1 | NP_001307414.1 | 10 | 1815 | |||
NM_001320487.1 | NP_001307416.1 | 10 | 1849 | |||
NM_025204.3 | NP_079480.2 | 10 | 1840 | |||
NR_135275.1 | 10 | 1811 | ||||
XM_011530713.2 | XP_011529015.2 | 10 | 1885 | |||
XM_017028944.1 | XP_016884433.1 | 10 | 1927 | |||
GenBank mRNA | AL449244.1 | - | 10 | 1821 | ||
AY203945.1 | - | 9 | 1887 | |||
BC093029.1 | - | 9 | 1913 | 30527560 |