Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184891.1 | NP_001171820.1 | 3 | 415 | ||
NM_001184892.1 | NP_001171821.1 | 3 | 1054 | |||
NM_025207.4 | NP_079483.3 | 2 | 859 | |||
NM_201398.2 | NP_958800.1 | 3 | 415 | |||
GenBank mRNA | AF218022.1 | - | 3 | 981 | ||
AF481877.1 | - | 2 | 167 | |||
AF520568.1 | - | 2 | 780 | |||
AK025979.1 | - | 3 | 343 | |||
AK090542.1 | - | 2 | 801 | |||
AK308243.1 | - | 3 | 349 | |||
BC011378.1 | - | 3 | 344 | 3855223 | ||
BC014012.2 | - | 1 | 134 | |||
BC021096.1 | - | 3 | 333 | 4891331 | ||
BC032323.1 | - | 3 | 350 | 5242727 | ||
BI919061.1 | - | 3 | 397 | |||
DC371856.1 | - | 3 | 416 | |||
DQ458779.1 | - | 2 | 458 |